Learn More
Restek™ FastPack Inlet Kits for Agilent GCs
Product Code. p-3181263
Encompass_Preferred
Click to view available options
Type:
Split w/Wool Inlet Maintenance Kit
Splitless Gooseneck Inlet Maintenance Kit
Splitless Inlet Maintenance Kit
Product Code. 13144492
Brand: Restek™ 21104
Only null left
Add to basket
Add to basket
Includes all parts needed for routine maintenance of injection ports and the entire system.
  • Economical - costs less than the sum of the individual parts
  • Factory sealed Mylar bag prevents contamination of products from weeks in the lab
  • Each bag includes five maintenance kits
  • Includes: inlet liner (choose from three popular styles), Viton O-ring, 0.8mm ID gold-plated inlet seal and washer, 11mm Thermolite septum
Specifications
Type Split w/Wool Inlet Maintenance Kit
Includes washer, septa and liner
For Use With Agilent GC