Learn More
Restek™ FastPack Inlet Kits for Agilent GCs
Click to view available options
Type:
Split w/Wool Inlet Maintenance Kit
Splitless Gooseneck Inlet Maintenance Kit
Splitless Inlet Maintenance Kit
Description
- Economical - costs less than the sum of the individual parts
- Factory sealed Mylar bag prevents contamination of products from weeks in the lab
- Each bag includes five maintenance kits
- Includes: inlet liner (choose from three popular styles), Viton O-ring, 0.8mm ID gold-plated inlet seal and washer, 11mm Thermolite septum
Specifications
Specifications
| Type | Splitless Inlet Maintenance Kit |
| Includes | washer, septa and liner |
| For Use With | Agilent GC |