missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Dual Vespel™ Ring Inlet Seals

Product Code. 13136962
Encompass_Preferred
Click to view available options
Diameter (Metric) Inner:
1.2 mm
This item is not returnable. View return policy

Product Code. 13136962

Brand: Restek™ 21248

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extend column lifetime by preventing oxygen from reaching the column with these inlet seals. Restek™ Dual Vespel™ Ring Inlet Seals eliminate the need for a washer and ensure very little torque is needed to make a leak-tight seal.

  • Washerless, Leak-Tight 1.2mm Seals for Agilent GCs
  • Two per pack
  • Does not require retightening of reducing nut after several oven cycles
  • Same price as the regular inlet seals with washers
  • Siltek treatment provides enhanced inertness versus stainless steel
  • Our patented dual Vespel ring inlet seal greatly improves injection port performance
  • It stays sealed, even after repeated temperature cycles, without retightening the reducing nut
  • This seal features two soft Vespel rings, one embedded in its top surface and the other embedded in its bottom surface
  • The rings will not harm the critical seal in the injector body, or any other surface, and are outside the sample flow path
  • Use a stainless steel seal for analyses of unreactive compounds
  • To reduce breakdown and adsorption of active compounds, use a gold-plated or Siltek-treated seal

Specifications

Diameter (Metric) Inner 1.2 mm
Material Siltek-treated
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.