Learn More
Restek™ Dual-Column Direct Injection Tee
Product Code. 13011770
Encompass_Preferred
Click to view available options
Type:
Tee
Tee Kit
Unit Size:
Each
Product Code. 13011770
Brand: Restek™ 20412
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For 0.25 in. packed column inlets.
  • Designed with a glass spiral to promote sample vaporization and provide even sample distribution to the two columns
  • Traps sample residue and minimizes the need for guard columns
  • Press-Tight taper in each outlet leg facilitates a perfect dead-volume-free connection to each analytical column (OD 0.4 to 0.8mm) and allows visual confirmation of the column connection
  • Includes all fittings and ferrules
Specifications
Type Tee Kit
For Use With Dual 0.32 and 0.53 mm I.D