missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Diskcover-47 Extraction Disk Holder

Product Code. 13129932
Encompass_Preferred
Click to view available options
Quantity:
6 Pack
Each
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13129932

Brand: Restek™ 24020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Compatible with most vacuum manifold systems that accept 0.125 in. male luer fittings.

  • Sample can be automatically introduced via 0.125 in. PTFE tubing or from the optional Diskcover-47 reservoir

Specifications

For Use With (Equipment) Most Vacuum Manifold Systems That Accept 0.12 in. Male Luer Fittings
Type Extraction Disk Holder
Quantity Each
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.