Learn More
Restek™ Detector Lamps
Product Code. p-3217634
Encompass_Preferred
Click to view available options
Type:
Deuterium Lamp
Lamp
For Use With (Equipment):
2487 Water Detector
996, 2996 Water Detector
Agilent VWD G1314 A HPLC System
Product Code. 10379622
Brand: Restek™ 25262
Used for waters detectors or for Agilent HPLC Systems
  • Standard lamps have nominal 1,000-hour life
Specifications
For Use With (Equipment) Agilent VWD G1314 A HPLC System
Includes 1 Chromatography Detector Lamp
Type Lamp