Learn More
Restek™ Detector Lamps
Product Code. 10379622
Encompass_Preferred
Change view
Click to view available options
Product Type:
Deuterium Lamp
Lamp
For Use With (Equipment):
2487 Water Detector
996, 2996 Water Detector
Agilent VWD G1314 A HPLC System
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type For Use With (Equipment)
10379622 Lamp Agilent VWD G1314 A HPLC System
10344783 Deuterium Lamp 996, 2996 Water Detector
10330495 Deuterium Lamp 2487 Water Detector
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10379622 Supplier Restek™ Supplier No. 25262
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Used for waters detectors or for Agilent HPLC Systems
  • Standard lamps have nominal 1,000-hour life
Specifications
Product Type Lamp
For Use With (Equipment) Agilent VWD G1314 A HPLC System
Includes 1 Chromatography Detector Lamp