Learn More
Restek™ Deactivated, Universal Press-Tight Connectors
Click to view available options
Packaging Quantity:
5
25
Description
- Deactivated fused quartz helps eliminate activity for complete inertness along the GC flow path
- Connect a guard column to an analytical column
- Repair a broken column
- Connect a column outlet to a transfer line
Specifications
Specifications
| Type | Connector |
| Packaging Quantity | 25 |
| For Use With | Fused Silica Capillary Column |