Learn More
Restek™ Cylinder Valve Wrench
Product Code. 13197032
Encompass_Preferred
Click to view available options
:
Cylinder valve wrench
Product Code. 13197032
Brand: Restek™ 21321
This specially designed wrench enables easy opening of cylinder valves that are fitted with a hand wheel.
  • Suitable for removing difficult cylinder caps
  • Easy-to-Use
Specifications
Type Cylinder Valve Wrench