Learn More
Restek™ Corporation EZ No-Vent™ GC Column-Mass Spectrometer Connector
Description
Includes
EZ No-Vent connecto, two 0.4mm ID adaptor ferrules for capillary column, two 0.4mm ID ferrules for transfer line, 100μm deactivated transfer line (3 ft.), column plug, column nut
EZ No-Vent GC column-mass spectrometer connector is simple and easy to use. A critical orifice in the EZ No-Vent connector minimizes the amount of oxygen allowed into the MS source, eliminating the need for purge gas. The EZ No-Vent connector easily attaches to the MS source without special tools or extra plumbing.
- Change GC-MS columns in minutes without venting - 100μmm transfer line maintains vacuum and eliminates the need to vent
- Easy to install and maintain; no special tools or plumbing required
- Gold-plated body for inertness
- High-temperature polyimide ferrules eliminate leaks at the problematic transfer line fitting
- Lower cost than other “no-vent” fittings
Specifications
Specifications
| Type | Connector |
| Diameter (Metric) Inner | 0.4 mm |
| Packaging Quantity | 1 Kit |
| Includes | Connector, Two 0.4 mm ID ferrules for capillary column, Two 0.4 mm ID ferrules for transfer line, 100 Micrometer deactivated transfer line 3 ft., Column plug and nμt |
| For Use With | Agilent 5971, 5972, 5973 GC/MS |