missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Copper Tubing

Product Code. 15155836
Encompass_Preferred
Click to view available options
Packaging Quantity:
50 ft.
This item is not returnable. View return policy

Product Code. 15155836

Brand: Restek™ 22628

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use this copper tubing for plumbing GC systems. Restek™ Copper Tubing is cleaned to remove residual organics.

  • Copper GC Tubing
  • 50 ft.

Specifications

Type Tubing
Diameter (English) Outer 1/8 in.
Diameter (English) Inner 0.065 in.
Packaging Quantity 50 ft.
Overall Length (English) 50 ft.
Material Copper
Pressure 2800 psig
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.