Learn More
Restek™ Ceramic Scoring Wafers
Click to view available options
Packaging Quantity:
5/Pk.
Description
- Four straight scoring edges for cutting fused silica tubing
- Four serrated edges for cutting MXT™ metal capillary columns
Specifications
Specifications
| Type | Scoring Wafer |
| Packaging Quantity | 5/Pk. |