Learn More
Restek™ Capillary Installation Gauge for Agilent GCs
Product Code. 13124572
Encompass_Preferred
Click to view available options
For Use With:
1/16 in. Ferrule Fittings
Agilent GC
Varian GCs, 0.0625 in. Ferrules
Unit Size:
Each
Product Code. 13124572
Brand: Restek™ 22335
Only null left
Add to basket
Add to basket
Seats graphite ferrules onto column for consistent installations.
  • Made from high-quality stainless steel
  • Prevents crushed column ends
  • For 1/16 in. ferrules and fittings
Part No. 21399
  • For Agilent GCs
Part No. 22335
  • For Bruker/Varian GCs
Specifications
Type Gauge
For Use With Varian GCs, 0.0625 in. Ferrules