Learn More
Restek™ Capillary Installation Gauge for Agilent GCs
Click to view available options
For Use With:
1/16 in. Ferrule Fittings
Agilent GC
Varian GCs, 0.0625 in. Ferrules
Unit Size:
Each
Description
- Made from high-quality stainless steel
- Prevents crushed column ends
- For 1/16 in. ferrules and fittings
- For Agilent GCs
- For Bruker/Varian GCs
Specifications
Specifications
| Type | Gauge |
| For Use With | 1/16 in. Ferrule Fittings |