Learn More
Restek™ Capillary Installation Gauge for Agilent MS
Click to view available options
Packaging Quantity:
1 Each
Description
- Made from high-quality stainless steel
- Seats ferrules onto column for consistent installations
Specifications
Specifications
| Type | Capillary Installation Gauge |
| For Use With (Application) | Seats Ferrules Onto Column for Consistent Installations |
| Packaging Quantity | 1 Each |
| For Use With | Agilent 5973 and 5975 MS |
| Material | Stainless Steel |