Learn More
Restek™ Capillary FID Jets for Agilent™ 5890/6890/6850 GC
Product Code. p-3181029
Encompass_Preferred
Click to view available options
Type:
High-Performance Siltek Treated
Standard
Packaging Quantity:
3 Pack
Each
Product Code. 12922599
Brand: Restek™ 20670
Only null left
Add to basket
Add to basket
Fluted tip to guide the capillary column into the jet
  • Threads specially coated for easy installation and removal
  • Standard version undergoes special processing to ensure the highest degree of cleanliness
  • High-performance version is Siltek™ treated to render extremely inert for use with active compounds
Specifications
Type Standard
Packaging Quantity Each
For Use With Agilent 5890/6890/6850