Learn More
Restek™ Capillary FID Jets for Agilent™ 5890/6890/6850 GC
Description
- Threads specially coated for easy installation and removal
- Standard version undergoes special processing to ensure the highest degree of cleanliness
- High-performance version is Siltek™ treated to render extremely inert for use with active compounds
Specifications
Specifications
| Type | High-Performance Siltek Treated |
| Packaging Quantity | Each |
| For Use With | Agilent 5890/6890/6850 |