Learn More
Restek™ Capillary Dedicated FID Jet for Agilent™ 6890/6850 GC
Fluted tip guides the capillary column into the jet
Brand: Restek™ 21683
Code : Z3
Additional Details : Weight : 1.00000kg
Description
- Threads specially coated for easy installation and removal
- Standard version undergoes special processing ensures the highest degree of cleanliness
- High-performance Siltek™ version is extremely inert for use with active compounds
Specifications
High-Performance Siltek Treated | |
3 Pack | |
Agilent 6890/6850 |