Learn More
Restek™ Capillary Dedicated FID Jet for Agilent™ 6890/6850 GC
Product Code. 10636213
Encompass_Preferred
Click to view available options
Type:
High-Performance Siltek Treated
Standard
Packaging Quantity:
3 Pack
Each
Product Code. 10636213
Brand: Restek™ 21682
Fluted tip guides the capillary column into the jet
  • Threads specially coated for easy installation and removal
  • Standard version undergoes special processing ensures the highest degree of cleanliness
  • High-performance Siltek™ version is extremely inert for use with active compounds
Specifications
Type Standard
Packaging Quantity 3 Pack
For Use With Agilent 6890/6850