Learn More
Restek™ Capillary Dedicated FID Jet for Agilent™ 6890/6850 GC
missing translation for 'orderingAttributeHoverText'
Type:
High-Performance Siltek Treated
Standard
Packaging Quantity:
3 Pack
Each
Description
- Threads specially coated for easy installation and removal
- Standard version undergoes special processing ensures the highest degree of cleanliness
- High-performance Siltek™ version is extremely inert for use with active compounds
Specifications
Specifications
| Type | High-Performance Siltek Treated |
| Packaging Quantity | 3 Pack |
| For Use With | Agilent 6890/6850 |