Learn More
Restek™ Capillary Column Rinsing Reservoir
Click to view available options
Type:
Rinsing Reservoir
Rinsing Reservoir Kit
Description
- 50mL rinsing reservoir equipped with 0.25 in. inlet and outlet connections
- Built-in fritted disk prevents particulate matter from contaminating the column
- Includes reservoir, pressure regulator, fittings, ferrules, and tubing
Order Info
Reservoir also available separately
Specifications
Specifications
| Type | Rinsing Reservoir Kit |
| Includes | Reservoir, pressure regulator, fittings, ferrules, and tubing |
| For Use With | Capillary Column |