Learn More
Restek™ Backpressure Regulator
Product Code. 12922569
Encompass_Preferred
Click to view available options
Operating Pressure:
0 to 60 psi
Unit Size:
Each
Product Code. 12922569
Brand: Restek™ 20635
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Maintains a constant upstream pressure and rapidly responds to catastrophic leaks.
  • For capillary GC inlet systems
  • Operating range of 0 to 60psig is sufficient to operate a 105m, 0.25mm ID column at its optimum flow rate
Specifications
For Use With (Equipment) Capillary GC Inlet System
Operating Pressure 0 to 60 psi
Type Backpressure Gas Regulator