Learn More
Restek™ Siltek FID Ignitor Castle for Agilent GC
Click to view available options
Type:
FID Ignitor Castle
Siltek FID Ignitor Castle
Description
For FID collector assembly kit for Agilent 5890, 6850, 6890, 7890 GCs.
Specifications
Specifications
| Type | FID Ignitor Castle |
| For Use With | Agilent 5890/6890/6850 |