missing translation for 'onlineSavingsMsg'
Learn More

Restek™ 11mm GC Snap-Top Cap with Septum

Product Code. 13199632
Encompass_Preferred
Click to view available options
Quantity:
100/Pk.
1000/Pk.
Material (Liner):
PTFE
PTFE/Butyl Rubber
PTFE/Silicone
This item is not returnable. View return policy

Product Code. 13199632

Brand: Restek™ 21732

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An effecitve alternative when there is no need to crimp caps

  • Snaps on any crimp-style vial
  • Clear
  • Polypropylene

Specifications

Material (Liner) PTFE/Butyl Rubber
Color Clear
Material Polypropylene
Quantity 1000/Pk.
Closure Size 11 mm
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.