missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ 1/8 to 1/4 in. Tube-end Union
To adapt 1/8 in. Super Clean base plate fittings to 1/4 in. tubing.
Brand: Restek™ 21833
This item is not returnable.
View return policy
Description
- Material: Brass
- Quantity: 5/Pk.
Specifications
Tube End Reducer | |
Inert Carrier Gas, Air, Hydrogen | |
0.12 in. Super-Clean™ base plate fittings to 0.25 in. tubing, use 0.12 in. to 0.25 in. tube end reducers | |
Brass |