Learn More
Restek™ 1/4-Inch Nut Driver
Product Code. 13154472
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
1 Ea.
Product Code. Packaging Quantity
13154472 1 Ea.
1 options
Product Code. 13154472 Supplier Restek™ Supplier No. 21076
Only null left
Add to basket
Add to basket
Ideal for changing Agilent 5890/6890/6850 FID jets
Securely grips jet for easy removal or installation.
Specifications
Type Nut Driver
Packaging Quantity 1 Ea.