Learn More
Restek™ 1/16 in. Tubing Cutting Pliers
Ideal for cutting 1/16 in. tubing
Brand:  Restek™ 20193
Encompass_Preferred
Product Code. 13051620
  • £106.48 / Each
Estimated Shipment: 04-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cut quickly, reducing distortion
  • Cut clean, eliminating need for deburring
Specifications
Specifications
Tubing Cutting Pliers
0.06 in.
0.16 cm
Ideal for Cutting 0.16 cm Tubing by Eliminating Tubing Distortion