Learn More
REST Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22242-1-AP-150UL
 View more versions of this product
Product Code. 16821785
  • £349.00 / 150µL
Description
Transcriptional repressor which binds neuron-restrictive silencer element (NRSE) and represses neuronal gene transcription in non-neuronal cells. Restricts the expression of neuronal genes by associating with two distinct corepressors, mSin3 and CoREST, which in turn recruit histone deacetylase to the promoters of REST-regulated genes. Mediates repression by recruiting the BHC complex at RE1/NRSE sites which acts by deacetylating and demethylating specific sites on histones, thereby acting as a chromatin modifier.
Specifications
Specifications
REST
Polyclonal
Unconjugated
REST
NRSF, REST, X2 box repressor, XBR
Rabbit
Antigen Affinity Chromatography
RUO
5978, 83618
-20°C
Liquid
Immunoprecipitation, Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O54963, Q13127
REST
REST Fusion Protein Ag17707
150 μL
Primary
Human, Rat