Learn More
REST Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-22242-20UL
Description
The REST antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human REST. This antibody recognizes human, rat antigen. The REST antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| REST | |
| 1000 μg/mL | |
| NRSF, REST, X2 box repressor, XBR | |
| 20 μL | |
| 5978 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Rat | |
| 150 kDa | |
| IgG |