missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus Nucleoprotein Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18802PE
This item is not returnable.
View return policy
Description
Respiratory Syncytial Virus Nucleoprotein Polyclonal antibody specifically detects Respiratory Syncytial Virus Nucleoprotein in Virus samples. It is validated for Western Blot
Specifications
| Respiratory Syncytial Virus Nucleoprotein | |
| Polyclonal | |
| Western Blot | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot | |
| PE | |
| PBS | |
| Produced in rabbits immunized with purified, recombinant Respiratory Syncytial Virus Nucleoprotein (Uniprot#: P03418; Met1-Leu391). | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction