missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus M2-1 Protein Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93589JF549
This item is not returnable.
View return policy
Description
Respiratory Syncytial Virus M2-1 Protein Polyclonal antibody specifically detects Respiratory Syncytial Virus M2-1 Protein in Virus samples. It is validated for Western Blot
Specifications
| Respiratory Syncytial Virus M2-1 Protein | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 549 | |
| Fusion glycoprotein F0, Fusion glycoprotein F1, Fusion glycoprotein F2, RS virus, RSV | |
| Reacts with residues 81-95 [LESYIGSINNITKQSA] of the human Respiratory Syncytial Virus M2 protein, which is a CTL epitope. 93% identity to bovine RSV M2. | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction