missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus Glycoprotein F Antibody (11-2-F3), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50412AF700
This item is not returnable.
View return policy
Description
Respiratory Syncytial Virus Glycoprotein F Monoclonal specifically detects Respiratory Syncytial Virus Glycoprotein F in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Respiratory Syncytial Virus Glycoprotein F | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Gradient-purified RSF-44 virus (subgroup A) UV inactivated for 20 minutes at 20C | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| This antibody reacts with the RS virus fusion glycoprotein F (also named VP70 or F0 protein), specifically to a subunit of the F protein, F1 (also named VGP48 or 48kDa), from both subgroups A and B of RS virus.This antibody has a high neutralising activity; 90% plaque neutralization titre of 102.2 (Gimenez et al 1996). This property makes it potentially useful for the preparation of reagents for prevention and/or therapeutic use (for example, vaccines or new-generation of man-made antibodies). The characterisation of the epitope of this antibody which induce high neutralising activity will be useful for the preparation of reagents (for example vaccine) to prevent RS virus infection. A RS virus vaccine is not yet available. A vaccine could be used to increase the maternal immunity to RS virus, which would then be passed to the newborn baby to provide passive immunity.The further characterisation of the epitope recognised by this antibody, which possesses a high neutralising activity, will be useful for the preparation of reagents to prevent RS virus infection. New-generation man-made antibodies, based on the 11-2-F3 antibody, could complement the effectiveness of polivizumab (Synagis), which is used to prevent but not treat established RS virus infections (Nature Biotechnology, 2013, 31, 952). | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 11-2-F3 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction