missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Respiratory Syncytial Virus fusion protein Antibody (RSV3216 (B016)), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64495JF646
This item is not returnable.
View return policy
Description
Respiratory Syncytial Virus fusion protein Monoclonal antibody specifically detects Respiratory Syncytial Virus fusion protein in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
| Respiratory Syncytial Virus fusion protein | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| RSV3216 (B016) | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| RSV fusion protein | |
| Bovine Respiratory Syncytial Virus fusion protein strains: 127, SNK and 9007. Human RSV strains: Long, Randall, 8/60, and A/2. | |
| 0.5 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction