Learn More
Moldex™ Respirator FFP2S
Click to view available options
Product Type:
Respirator FFP2S
Unit Size:
Pack of 10
Description
Moldex Smart Special FFP2 NR D welding valved dust mask. The activated layer of charcoal built-into this mask offers relief from low levels of gases and vapours, often found in conjunction with particulate hazards. FFP2 NR D + Nuisance acid & organic gas relief.
Specifications
Specifications
| Disposable | Single-use |
| Product Type | Respirator FFP2S |
| Quantity | 10/Pk. |