Learn More
uvex Respirator Face Mask
Product Code. 15395792 Shop All Uvex Products
Click to view available options
Model:
3220 silv-Air Classic
Classic cup style FFP3
silv-Air c 2200 FFP2
silv-Air c 3310 FFP3
Unit Size:
Each
Pack of 15
Pack of 20
Product Code. 15395792
Brand: uvex 8733220
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Breathing protection preformed mask
  • Preformed mask
  • Labelled for use with high levels of dust exposure (D)
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 149:2001 + A1:2009
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type FFP2
Material Polypropylene, Polyester
Product Type Particulate Respirator
Size Regular
With Exhalation Valve Yes
Filter Efficiency FFP2
Model 3220 silv-Air Classic
Style Flat fold