missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RESP18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561108
This item is not returnable.
View return policy
Description
Immunogen sequence: PTKTTESPLA KVNRDQCFTS EVVSKALKQE VANPVKITYR CSYGGLDMMQ APGPSKEEII YKIMRLLWAT S Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 28%.
Specifications
| RESP18 | |
| Polyclonal | |
| Unconjugated | |
| RESP18 | |
| regulated endocrine specific protein 18; regulated endocrine-specific protein 18; regulated endocrine-specific protein 18 homolog; RESP18 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 389075 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5W5W9 | |
| RESP18 | |
| Recombinant protein corresponding to Human RESP18. Recombinant protein control fragment (Product #RP-96212). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction