missing translation for 'onlineSavingsMsg'
Learn More

Resorufin, TRC

Product Code. 30425199
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425199 250 mg 250mg
30414818 500 mg -
30413624 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30425199 Supplier Toronto Research Chemicals Supplier No. TRCR146515250MG
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 635-78-9
Molecular Formula C12H7NO3
Molecular Weight (g/mol) 213.19
Synonym 7-Hydroxy-3H-phenoxazin-3-one, 7-Hydroxyphenoxaz-3-one, 7-Hydroxyphenoxazin-3-one, NSC 12097, Resorufin, NSC 12097, Resofurin, Resorufine
IUPAC Name 7-hydroxy-3H-phenoxazin-3-one
SMILES O=C(C=C1)C=C2C1=NC3=CC=C(O)C=C3O2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H7NO3/c14-7-1-3-9-11(5-7)16-12-6-8(15)2-4-10(12)13-9/h1-6,14H
Formula Weight 213.04
Chemical Name or Material Resorufin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.