missing translation for 'onlineSavingsMsg'
Learn More

Resorcinol monoacetate, MedChemExpress

Product Code. 16488486
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488486 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488486 Supplier Medchem Express Supplier No. HYB0894
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Resorcinol monoacetate (Acetylresorcinol) is an antiseptic and a disinfectant, is a chemical intermediate for the production of many other pharmaceuticals, and has the potential for acne, seborrheic dermatitis, eczema, psoriasis, and other skin disorders research.

Resorcinol monoacetate (Acetylresorcinol) is an antiseptic and a disinfectant, is a chemical intermediate for the production of many other pharmaceuticals, and has the potential for acne, seborrheic dermatitis, eczema, psoriasis, and other skin disorders research.

Specifications

Chemical Name or Material Resorcinol monoacetate
CAS 102-29-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C8H8O3
Quantity 100 mg
Synonym Acetylresorcinol Resorcin monoacetate
Solubility Information DMSO : ≥ 43 mg/mL (282.62 mM)
SMILES OC1=CC(OC(C)=O)=CC=C1
Molecular Weight (g/mol) 152.15
Formula Weight 152.15
Percent Purity 96.22%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.