missing translation for 'onlineSavingsMsg'
Learn More

Resorcinol Crystalline MP Biomedicals

Product Code. 11406192 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406192 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11406192 Supplier MP Biomedicals Supplier No. 0210280825
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 108-46-3
Density 3.79 (air = 1)
Molecular Formula C6H6O2
Quantity 25 g
Synonym resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
InChI Key GHMLBKRAJCXXBS-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)O)O
IUPAC Name benzene-1,3-diol
Molecular Weight (g/mol) 110.112
PubChem CID 5054
ChEBI CHEBI:27810
Formula Weight 110.1
Physical Form Crystalline
Chemical Name or Material Resorcinol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.