missing translation for 'onlineSavingsMsg'
Learn More

Resorcinol, ACS, 99.0-100.5%

Product Code. p-7050233
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11445160

Brand: Thermo Scientific Alfa Aesar 036248.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-46-3 | C6H6O2 | 110.112 g/mol

Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-46-3
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00002269
InChI Key GHMLBKRAJCXXBS-UHFFFAOYSA-N
Synonym resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
PubChem CID 5054
ChEBI CHEBI:27810
IUPAC Name benzene-1,3-diol
SMILES C1=CC(=CC(=C1)O)O

Specifications

Melting Point 109°C to 112°C
Color White to Cream
Density 1.272 g/mL
pH 4 to 6
Boiling Point 280°C to 281°C
Flash Point 171°C (339°F)
Assay Percent Range 99.0 to 100.5%
Odor Phenol-like
Quantity 25 g
UN Number UN2876
Beilstein 906905
Sensitivity Air and light sensitive
Merck Index 14,8155
Solubility Information Soluble in water,alcohol,ether and glycerol.
Formula Weight 110.11
Percent Purity 99.0 to 100.5%
Grade ACS
Physical Form Crystalline
Chemical Name or Material Resorcinol, For analysis ACS
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Resorcinol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Resorcinol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.