Learn More
Biochrom Resolution Software, CFR for BioDrop UV-Vis Spectrophotometers
Product Code. 15871232
Click to view available options
Type:
Software
Unit Size:
Each
Product Code. 15871232
Brand: Biochrom 80300670
Resolution CFR Software offers the ability to set up different users as part of defined groups, audit trail, electronic signature and many other features. For labs that need full 21 CFR part 11 compliance and compatible with Biochrom's WPA, Libra, and BioDrop spectrophotometers.
  • Quick Read and quick scan
  • Wavelength scanning, kinetics, qunatitative calibration curves
  • Validation
  • Medthos developer
  • Life science methods
  • Accessory control
Specifications
Description Resolution Software, CFR for BioDrop UV-Vis Spectrophotometers
Type Software
Disposable Reusable
For Use With (Equipment) Biochrom Spectrophotometers