missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Resistin Recombinant Rabbit Monoclonal Antibody (002), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£74.50 - £359.00
Specifications
| Antigen | Resistin |
|---|---|
| Clone | 002 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30019711
|
Invitrogen™
MA552393 |
20 μL |
£74.50
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019833
|
Invitrogen™
MA552392 |
100 μL |
£359.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Resistin Recombinant Monoclonal antibody specifically detects Resistin in Mouse samples. It is validated for ELISASpecifications
| Resistin | |
| 1 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| Adipose tissue specific secretory factor; adipose tissue-specific secretory factor; Adipose-specific cysteine-rich secreted protein A12-alpha; ADSF; C/EBP epsilon regulated myeloid specific secreted cysteine rich protein precursor 1; C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich protein precursor 1; c/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein; cysteine-r; Cysteine-rich secreted protein A12-alpha-like 2; cysteine-rich secreted protein FIZZ3; dominant inhibitory adipocyte-specific secretory factor; dri; FIZZ3; found in inflammatory zone 3; HXCP1; MGC126603; MGC126609; Resistin; resistin delta2; RETN; RETN1; RSN; RSTN; UNQ407/PRO1199; XCP1; Xcp4 | |
| Recombinant Mouse Resistin Protein. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| RETN |
| 002 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Q99P87 | |
| 57264 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title