missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Resistin Mouse anti-Human, Clone: RETN/3331, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07242-100ug
This item is not returnable.
View return policy
Description
Resistin Monoclonal specifically detects Resistin in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Resistin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Adipose tissue-specific secretory factor, ADSFMGC126609, C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 1, C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein, Cysteine-rich secreted protein A12-alpha-like 2, Cysteine-rich secreted protein FIZZ3, FIZZ3MGC126603, found in inflammatory zone 3, HXCP1, resistin, RETN1, RSTNXCP1 | |
| Recombinant fragment of human Resistin protein (around aa 19-108) (exact sequence is proprietary) | |
| 100 μg | |
| Diabetes Research | |
| 56729 | |
| Store at 4°C. | |
| IgG2b κ |
| ELISA, Peptide Array | |
| RETN/3331 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction