missing translation for 'onlineSavingsMsg'
Learn More

Resiquimod, MedChemExpress

Product Code. 16431987
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16431987

Brand: Medchem Express HY13740

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Resiquimod is a Toll-like receptor 7 and 8 (TLR7/TLR8) agonist that induces the upregulation of cytokines such as TNF-α, IL-6 and IFN-α.

Resiquimod is a Toll-like receptor 7 and 8 (TLR7/TLR8) agonist that induces the upregulation of cytokines such as TNF-α, IL-6 and IFN-α.

Specifications

Chemical Name or Material Resiquimod
CAS 144875-48-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H22N4O2
Quantity 50 mg
Synonym R848 S28463
Solubility Information DMF : 50 mg/mL (159.04 mM; Need ultrasonic) ∣DMSO : ≥ 30 mg/mL (95.43 mM) ∣Methanol : 25 mg/mL (79.52 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.32 mM; Need ultrasonic)
SMILES OC(C)(C)CN1C(COCC)=NC2=C1C3=CC=CC=C3N=C2N
Molecular Weight (g/mol) 314.38
Formula Weight 314.38
Percent Purity 99.39%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.