Learn More
Fisherbrand™ Multi-Channel Reservoirs
Support full sample recovery with tapered reservoirs. Fisherbrand™ Multi-Channel Reservoirs are designed for superior handling with thicker, crush-resistant walls. Disposable reservoirs are available in two sizes of sterile or nonsterile options, plus a clear PVC option for better sample visibility.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.54000kg
Description
- Thick walls of white polystyrene or clear PVC, with crush-resistant turrets at the corners
- Tapered reservoir bottoms and easy-pour spouts to aid full sample recovery
- Stackable, with tabs to prevent stacked reservoirs from sticking
- 55mL and 100mL volumes, both with 25mL graduation marks
- Sterile versions available
- Gamma-irradiated in an ISO-certified facility
- Individually sealed in Sani-Tear HDPE packets
Specifications
White | |
145 x 85 x 25mm | |
Non-sterile | |
100/Pk. |
55 mL | |
Polystyrene | |
Reagent Reservoir | |
Single-use |