missing translation for 'onlineSavingsMsg'
Learn More

Reserpine, 98%

Product Code. 10737573 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10737573 1 g 1g
10478480 5 g 5g
10381004 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10737573 Supplier Thermo Scientific Acros Supplier No. 132280010
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50-55-5 | C33H40N2O9 | 608.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-55-5
Molecular Formula C33H40N2O9
Molecular Weight (g/mol) 608.69
InChI Key QEVHRUUCFGRFIF-MDEJGZGSSA-N
IUPAC Name methyl (1R,15S,17R,18R,19S,20S)-6,18-dimethoxy-17-(3,4,5-trimethoxybenzoyloxy)-3,13-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8-tetraene-19-carboxylate
SMILES CO[C@H]1[C@@H](C[C@@H]2CN3CCC4=C(NC5=CC(OC)=CC=C45)[C@H]3C[C@@H]2[C@@H]1C(=O)OC)OC(=O)C1=CC(OC)=C(OC)C(OC)=C1

Specifications

Melting Point 265.0°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 8265
Specific Rotation -126.00
Formula Weight 608.69
Chemical Name or Material Reserpine
compliance-icons compliance-icons
Product Identifier
  • Reserpine
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1A
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H351-Suspected of causing cancer.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.