missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ resDNASEQ™ Quantitative NS0 DNA Kits

Product Code. 30000686 Shop All Applied Biosystems Products
Change view
Click to view available options
Includes:
Assay Kit and Sample Preparation
Assay Kit only
Unit Size:
100 reactions
Product Code. Includes unitSize
30000686 Assay Kit only 100 reactions
30000692 Assay Kit and Sample Preparation 100 reactions
2 options
This item is not returnable. View return policy

Product Code. 30000686

Brand: Applied Biosystems™ 4458441

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The resDNASEQ™ Quantitative NS0 DNA Kit is a quantitative PCR (qPCR)-based system for the detection of host cell DNA from NS0 (murine myeloma) cells, an expression system commonly used for the production of vaccines.

The resDNASEQ™ Quantitative NS0 DNA Kit is a quantitative PCR (qPCR)-based system for the detection of host cell DNA from NS0 (murine myeloma) cells, an expression system commonly used for the production of vaccines. Reliable and rapid, the resDNASEQ™ system enables sensitive (as low as 1.5 pg DNA/mL test sample) and specific quantitation of NS0 cell DNA. This performance helps ensure a high degree of confidence in quantitation data obtained from a broad range of sample types, from in-process samples to final product, whether the sample contains high molecular weight or sheared DNA.

Features of the resDNASEQ Quantitative NS0 DNA Kit include:
• Highly sensitive quantitation using proven TaqMan™ real-time qPCR technology
• Consistent performance across the expected range of DNA fragment sizes
• Integrated system includes sample preparation kit, master mix, TaqMan™ primer/probe mix, and NS0 genomic DNA standard

This kit is available with or without sample preparation reagents in the form of a PrepSEQ™ Residual DNA Sample Preparation Kit, which is optimized for highly efficient residual DNA recovery from complex mixtures of proteins, buffers, and salts.

The resDNASEQ™ Quantitative NS0 DNA Kit is designed to provide highly sensitive detection of NS0 DNA, allowing you to use small sample volumes to generate accurate results. The broad linear range of TaqMan™ real-time qPCR technology allows you to test samples with variable levels of NS0 DNA in the same assay, such as in-process samples with higher amounts of DNA or bulk drug substance with very low amounts.

Easy to use, results in under 4 hours
The easy workflow of the resDNASEQ™ Quantitative NS0 DNA Kit consists of sample preparation, assay setup, and instrument run and data analysis—all of which you can perform in under 4 hours. Start by extracting DNA from your in-process purification and drug substance samples. Then perform qPCR analysis to compare the amount of NS0 DNA in your test samples to a standard curve generated with known amounts of purified NS0 genomic DNA standard. The resDNASEQ™ system includes almost everything you need to carry out these steps, from sample preparation kit to master mix to TaqMan™ primer/probe mix to NS0 genomic DNA standard, as well as step-by-step instructions.

Specific to NS0 DNA, no cross-reactivity with unrelated DNA
The target of the assay is highly specific—it only detects a NS0-specific region of a multicopy genetic element. We selected this region using extensive bioinformatic analysis of multiple related and unrelated species.

Consistent performance even with fragmented DNA
For accurate quantitation of residual NS0 DNA, assay results must be unaffected by the size of the DNA molecules present in the test sample. With the resDNASEQ™ system you get consistent assay performance across a range of DNA fragment sizes.

Specifications

Content And Storage resDNASEQ NS0 DNA Control
• NS0 DNA Control, 30 ng/μL, 40 μL, store at –15 to –25°C
• DNA Dilution Buffer (DDB), 7 mL, store at –15 to –25°C before first use and 2−8°C after first use

resDNASEQ NS0 DNA Real-Time PCR Reagents
• Environmental Master Mix (2X), 2 x 0.75 mL, store at –15 to –25°C before first use and 2−8°C after first use, protect from light
• NS0 DNA Assay Mix (10X), 300 μL, store at –15 to –25°C, protect from light
• Negative Control (water), 1.0 mL, store at –15 to –25°C before first use and 2−8°C after first use

Detection Method Primer-probe
Format Kit
PCR Method qPCR
Host Cell NS0
Label or Dye FAM
Product Line resDNASEQ
Quantity 100 reactions
Shipping Condition Dry Ice
No. of Reactions 100 Reactions
Includes Assay Kit only
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.