missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ resDNASEQ™ Quantitative MDCK DNA Kits

Product Code. 30000679 Shop All Applied Biosystems Products
Click to view available options
Includes:
Assay Kit and Sample Preparation
Assay Kit only
Unit Size:
100 reactions
This item is not returnable. View return policy

Product Code. 30000679

Brand: Applied Biosystems™ 4464335

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The resDNASEQ™ Quantitative MDCK DNA Kit is a quantitative PCR (qPCR)-based system for the detection of host cell DNA from MDCK (canine kidney) cells, an expression system commonly used for the production of vaccines.

The resDNASEQ™ Quantitative MDCK DNA Kit is a quantitative PCR (qPCR)-based system for the detection of host cell DNA from MDCK (canine kidney) cells, an expression system commonly used for the production of vaccines. Reliable and rapid, the resDNASEQ™ system enables sensitive (as low as 1.5 pg DNA/mL test sample) and specific quantitation of MDCK cell DNA. This performance helps ensure a high degree of confidence in quantitation data obtained from a broad range of sample types, from in-process samples to final product, if the sample contains high molecular weight or sheared DNA.

Features of the resDNASEQ Quantitative MDCK DNA Kit include:
• Highly sensitive quantitation using proven TaqMan™ real-time qPCR technology
• Consistent performance across the expected range of DNA fragment sizes
• Includes master mix, TaqMan™ primer/probe mix, and MDCK genomic DNA standard

This kit is available with or without sample preparation reagents in the form of a PrepSEQ™ Residual DNA Sample Preparation Kit, which is optimized for highly efficient residual DNA recovery from complex mixtures of proteins, buffers, and salts.

The resDNASEQ™ Quantitative MDCK DNA Kit is designed to provide highly sensitive detection of MDCK DNA, allowing you to use small sample volumes to generate accurate results. The broad linear range of TaqMan™ real-time qPCR technology allows you to test samples with variable levels of MDCK DNA in the same assay, such as in-process samples with higher amounts of DNA or bulk drug substance with very low amounts.

Easy to use
With the resDNASEQ™ Quantitative MDCK DNA Kit you perform qPCR analysis to compare the amount of MDCK DNA in your test samples to a standard curve generated with known amounts of purified MDCK genomic DNA standard. The resDNASEQ™ kit includes almost everything you need to carry out these steps, from master mix to TaqMan™ primer/probe mix to MDCK genomic DNA standard, as well as step-by-step instructions.

Specific to MDCK DNA, no cross-reactivity with unrelated DNA
The target of the assay is highly specific—it only detects a MDCK-specific region of a multicopy genetic element. We selected this region using extensive bioinformatic analysis of multiple related and unrelated species.

Consistent performance even with fragmented DNA
For accurate quantitation of residual MDCK DNA, assay results must be unaffected by the size of the DNA molecules present in the test sample. With the resDNASEQ™ system you get consistent assay performance across a range of DNA fragment sizes.

Specifications

Content And Storage resDNASEQ MDCK DNA Control
• MDCK DNA Control, 30 ng/μL, 40 μL, store at –15 to –25°C
• DNA Dilution Buffer (DDB), 7 mL, store at –15 to –25°C before first use and 2−8°C after first use

resDNASEQ MDCK DNA Real-Time PCR Reagents
• Environmental Master Mix (2X), 2 x 0.75 mL, store at –15 to –25°C before first use and 2−8°C after first use, protect from light
• MDCK DNA Assay Mix (10X), 300 μL, store at –15 to –25°C, protect from light
• Negative Control (water), 1.0 mL, store at –15 to –25°C before first use and 2−8°C after first use

Detection Method Primer-probe
Format Kit
PCR Method qPCR
Host Cell MDCK
Label or Dye FAM
Product Line resDNASEQ
Quantity 100 reactions
Shipping Condition Dry Ice
No. of Reactions 100 Reactions
Includes Assay Kit only
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.