missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ resDNASEQ™ Quantitative E1A DNA Fragment Length Kits

Product Code. 18775515 Shop All Applied Biosystems Products
Click to view available options
Includes:
Assay Kit and Sample Preparation
Assay Kit only
Unit Size:
100 reactions
Each
This item is not returnable. View return policy

Product Code. 18775515

Brand: Applied Biosystems™ A51969

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The resDNASEQ Quantitative E1A DNA Fragment Length Kit is a quantitative PCR (qPCR)-based system that measures and quantitates the fragment lengths of residual host-cell DNA by targeting the E1A gene in HEK 293 cell lines.

The resDNASEQ Quantitative E1A DNA Fragment Length Kit is a quantitative PCR (qPCR)-based system that measures and quantitates the fragment lengths of residual host-cell DNA by targeting the E1A gene in HEK 293 cell lines, which are predominantly used in the development of gene therapy, cell-based vaccines, and similar biotherapeutics. Reliable and rapid, the resDNASEQ kit enables sensitive and specific quantitation of different fragment lengths of the E1A gene within the residual host-cell DNA. This performance helps ensure a high degree of confidence in the obtained quantitation data.

Features of the resDNASEQ Quantitative E1A DNA Fragment Length Kit include:
• Highly sensitive quantitation using proven TaqMan real-time qPCR technology
• Consistent performance across the expected range of DNA fragment sizes
• Includes master mix, three TaqMan primer/probe mixes, and a single E1A plasmid DNA standard

This kit is available with or without sample preparation reagents in the form of a PrepSEQ Residual DNA Sample Preparation Kit, which is optimized for highly efficient residual DNA recovery from complex mixtures of proteins, buffers, and salts.

Integrated workflow solution
The resDNASEQ Quantitative E1A DNA Fragment Length Kit is part of an integrated workflow for impurity and contaminant testing during biopharmaceutical manufacturing. The kit is compatible with the PrepSEQ Residual DNA Sample Preparation Kit, which offers both a manual and automated sample preparation workflow. The resDNASEQ kit has been validated on the Applied Biosystems 7500 Fast Real-Time PCR System and the QuantStudio 5 Real-Time PCR System. Data analysis is streamlined using AccuSEQ Real-Time PCR Detection Software that provides accurate quantitation and security, audit, and e-signature capabilities to help enable 21 CFR Pt 11 compliance.

Easy to use
With the resDNASEQ Quantitative E1A DNA Fragment Length Kit, you perform qPCR analysis to quantitate the copy numbers of three different fragment size ranges in your test samples and compare that against standard curves generated with known amounts of specific fragments of the E1A DNA standard. The resDNASEQ kit includes almost everything you need to carry out these steps, from master mix to TaqMan primer/probe mix to E1A DNA standards, as well as step-by-step instructions.

Consistent performance
For accurate quantitation of fragments of three different lengths of E1A DNA, assay results must be unaffected by the size of non-specific DNA present in the test sample. With the resDNASEQ system you get consistent assay performance across the three E1A fragment sizes.

Specifications

Content And Storage resDNASEQ E1A DNA Control
• E1A DNA Control, 6.30 x 108 copies/μL, 0.12 mL, store at –25 to –15°C
• DNA Dilution Buffer (DDB), 7 mL, store at –25 to –15°C before first use and 2−8°C after first use

resDNASEQ E1A DNA Real-Time PCR Reagents
• Environmental Master Mix (2X), 5 mL, store at –15 to –25°C before first use and 2−8°C after first use, protect from light
• E1A DNA Short Fragment Assay Mix, 1 x 400 μL, store at –15 to –25°C, protect from light
• E1A DNA Medium Fragment Assay Mix, 1 x 660 μL,  store at –15 to –25°C, protect from light
• E1A DNA Long Fragment Assay, 1 x 400 μL, store at –15 to –25°C, protect from light
• Negative Control (water), 3 x 1.0 mL, store at –15 to –25°C before first use and 2−8°C after first use

Detection Method Primer-probe
Format Kit
PCR Method qPCR
Host Cell HEK
Label or Dye FAM
Product Line resDNASEQ
Quantity 100 reactions
Shipping Condition Dry Ice
No. of Reactions 100 Reactions
Includes Assay Kit only
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.