missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Resazurin sodium salt

Product Code. 11474687
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474687 5 g 5g
11464687 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474687 Supplier Thermo Scientific Chemicals Supplier No. B21187.06
  • ONLINE EXCLUSIVE PRICE
    £79.10
    £64.65 / 5g
    Save £14.45
    18% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

As indicator pH 3.8 (orange) to 6.5 (dark violet) | CAS: 62758-13-8 | C12H6NNaO4 | 251.173 g/mol

As indicator pH 3.8 (orange) to 6.5 (dark violet)Resazurin sodium salt acts as a blue non-fluorescent dye used especially in oxidation-reduction indicator of resazurin test for bacteria. It is employed as a fluorogenic oxidation-reduction indicator in a variety of cells, yeast and eukaryotes by flow cytometry, fluorescence microscopy and high-throughput screening. Further, it is used to characterize the metabolic activity of mitochnodria. It is also used to assay L-Glutamate and to measure the aerobic biodegradation of organic matter exists in effluents. In addition to this, it acts as an indicator for cell viability in mammalian cell cultures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As indicator pH 3.8 (orange) to 6.5 (dark violet)Resazurin sodium salt acts as a blue non-fluorescent dye used especially in oxidation-reduction indicator of resazurin test for bacteria. It is employed as a fluorogenic oxidation-reduction indicator in a variety of cells, yeast and eukaryotes by flow cytometry, fluorescence microscopy and high-throughput screening. Further, it is used to characterize the metabolic activity of mitochnodria. It is also used to assay L-Glutamate and to measure the aerobic biodegradation of organic matter exists in effluents. In addition to this, it acts as an indicator for cell viability in mammalian cell cultures.

Solubility
Soluble in water, dimethylsulfoxide and ammonium hydroxide.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62758-13-8
Molecular Formula C12H6NNaO4
Molecular Weight (g/mol) 251.173
MDL Number MFCD00005036
InChI Key IVGPGQSSDLDOLH-UHFFFAOYSA-M
PubChem CID 112939
IUPAC Name sodium;10-oxido-7-oxophenoxazin-10-ium-3-olate
SMILES C1=CC2=C(C=C1[O-])OC3=CC(=O)C=CC3=[N+]2[O-].[Na+]

Specifications

Beilstein 3794971
Solubility Information Soluble in water,dimethylsulfoxide and ammonium hydroxide.
Quantity 5 g
Formula Weight 251.18
Chemical Name or Material Resazurin
compliance-icons
Product Identifier
  • Resazurin sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Resazurin sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.